Product Details
- SNP ID
-
rs114241480
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:97775784 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATGGCCAAGTCCGCCCCGCCCCCT[C/T]CCCGTCCCCGCCGCTGCAGCGGTCG
- Phenotype
-
MIM: 613296
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LAPTM4B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2449510] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LAPTM4B
- Gene Name
- lysosomal protein transmembrane 4 beta
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018407.4 |
204 |
Silent Mutation |
CTC,CTT |
L16L |
NP_060877.3 |
View Full Product Details