Product Details

SNP ID
rs114550597
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:17160123 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACAGATCCCTTATGGCTGCTGCT[G/T]TGTTAACTTCGCTTGTTGAAGAAGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC105379298 PubMed Links

Gene Details

Gene
LOC105379298
Gene Name
uncharacterized LOC105379298
There are no transcripts associated with this gene.

Gene
ZDHHC2
Gene Name
zinc finger DHHC-type containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016353.4 Intron NP_057437.1
XM_011544544.2 Intron XP_011542846.1
XM_011544545.2 Intron XP_011542847.1
XM_011544546.2 Intron XP_011542848.1
XM_011544547.2 Intron XP_011542849.1
XM_011544548.2 Intron XP_011542850.1
XM_011544549.2 Intron XP_011542851.1

View Full Product Details