Product Details

SNP ID
rs114641823
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:92885312 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCCATATACTCCTTAGATATTT[C/T]CCAAGGATTTCTTCTCTTGGCTAGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRIQK PubMed Links

Gene Details

Gene
TRIQK
Gene Name
triple QxxK/R motif containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171795.1 2044 UTR 3 NP_001165266.1
NM_001171796.1 2044 UTR 3 NP_001165267.1
NM_001171797.1 2044 UTR 3 NP_001165268.1
NM_001171798.1 2044 UTR 3 NP_001165269.1
NM_001171799.1 2044 UTR 3 NP_001165270.1
NM_001191035.1 2044 UTR 3 NP_001177964.1
NM_001191036.1 2044 UTR 3 NP_001177965.1
XM_017013347.1 2044 UTR 3 XP_016868836.1
XM_017013348.1 2044 UTR 3 XP_016868837.1
XM_017013349.1 2044 UTR 3 XP_016868838.1
XM_017013350.1 2044 UTR 3 XP_016868839.1

View Full Product Details