Product Details

SNP ID
rs115055201
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:145003404 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGACCTATTGCGGTGGAGCTGGAG[G/T]TCGAGAACCAACCACTCGGTGCTGA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TMED10P1 PubMed Links

Gene Details

Gene
TMED10P1
Gene Name
transmembrane p24 trafficking protein 10 pseudogene 1
There are no transcripts associated with this gene.

Gene
ZNF252P
Gene Name
zinc finger protein 252, pseudogene
There are no transcripts associated with this gene.

Gene
ZNF252P-AS1
Gene Name
ZNF252P antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details