Product Details

SNP ID
rs115103902
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144481320 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCAGGCAATAGTGATCGCTTTCT[A/G]TTACTGTAGGTTTGGCGTCCCTTTT
Phenotype
MIM: 603621 MIM: 615216 MIM: 609172
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FOXH1 PubMed Links

Gene Details

Gene
FOXH1
Gene Name
forkhead box H1
There are no transcripts associated with this gene.

Gene
KIFC2
Gene Name
kinesin family member C2
There are no transcripts associated with this gene.

Gene
PPP1R16A
Gene Name
protein phosphatase 1 regulatory subunit 16A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005272350.2 Intron XP_005272407.1
XM_011517349.2 Intron XP_011515651.1
XM_017013912.1 Intron XP_016869401.1
XM_017013913.1 Intron XP_016869402.1
XM_017013914.1 Intron XP_016869403.1
XM_017013915.1 Intron XP_016869404.1

View Full Product Details