Product Details

SNP ID
rs115414296
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:119841746 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCAACTCAAGTATCATTCACCTA[A/G]TAATTAATCAACTGTTGAAGTAAGG
Phenotype
MIM: 613203 MIM: 604912
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DSCC1 PubMed Links

Gene Details

Gene
DSCC1
Gene Name
DNA replication and sister chromatid cohesion 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024094.2 Intron NP_076999.2
XM_005251065.3 Intron XP_005251122.1
Gene
TAF2
Gene Name
TATA-box binding protein associated factor 2
There are no transcripts associated with this gene.

View Full Product Details