Product Details

SNP ID
rs115422395
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:380574 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACACTCTGCTCTCCTGTGCCAAG[C/T]GTCAATATGGATTTTTGATGAAATT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM87A PubMed Links

Gene Details

Gene
FAM87A
Gene Name
family with sequence similarity 87 member A
There are no transcripts associated with this gene.

View Full Product Details