Product Details

SNP ID
rs115515473
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:22148443 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCAGGGCCTGCACCTCCGAGAAG[C/T]GGGTACCCTCCCAGCGCAGGATCTT
Phenotype
MIM: 608302 MIM: 609349
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LGI3 PubMed Links

Gene Details

Gene
LGI3
Gene Name
leucine rich repeat LGI family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139278.2 1653 Missense Mutation CAC,CGC H455R NP_644807.1
Gene
REEP4
Gene Name
receptor accessory protein 4
There are no transcripts associated with this gene.

View Full Product Details