Product Details

SNP ID
rs116019502
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:242991 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTTCAAGATGTGATGTTGGAGAA[C/T]ATCAGTCATCTGGTCTCTATTGGTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF596 PubMed Links

Gene Details

Gene
ZNF596
Gene Name
zinc finger protein 596
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042415.2 405 Silent Mutation AAC,AAT N39N NP_001035880.1
NM_001042416.2 405 Silent Mutation AAC,AAT N39N NP_001035881.1
NM_001287254.1 405 Silent Mutation AAC,AAT N39N NP_001274183.1
NM_001287255.1 405 Silent Mutation AAC,AAT N39N NP_001274184.1
NM_001287256.1 405 Silent Mutation AAC,AAT N39N NP_001274185.1
NM_001287399.1 405 UTR 5 NP_001274328.1
NM_173539.3 405 Silent Mutation AAC,AAT N39N NP_775810.2
XM_011534744.1 405 Silent Mutation AAC,AAT N4N XP_011533046.1
XM_017013166.1 405 Silent Mutation AAC,AAT N39N XP_016868655.1

View Full Product Details