Product Details

SNP ID
rs116984236
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:141207426 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATAGAAGAGAGTCATGAGCGACA[C/T]GACACTTCCTTACGACCTCTCTCTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC45A4 PubMed Links

Gene Details

Gene
SLC45A4
Gene Name
solute carrier family 45 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080431.2 6973 UTR 3 NP_001073900.1
NM_001286646.1 6973 UTR 3 NP_001273575.1
NM_001286648.1 6973 Intron NP_001273577.1
XM_011517169.2 6973 UTR 3 XP_011515471.1
XM_011517170.2 6973 UTR 3 XP_011515472.1
XM_011517171.2 6973 UTR 3 XP_011515473.1
XM_011517172.2 6973 UTR 3 XP_011515474.1
XM_011517173.2 6973 Intron XP_011515475.1
XM_011517174.2 6973 UTR 3 XP_011515476.1
XM_011517176.2 6973 UTR 3 XP_011515478.1
XM_011517177.2 6973 UTR 3 XP_011515479.1
XM_011517178.2 6973 UTR 3 XP_011515480.1
XM_011517179.2 6973 UTR 3 XP_011515481.1
XM_017013648.1 6973 UTR 3 XP_016869137.1
XM_017013649.1 6973 UTR 3 XP_016869138.1
XM_017013650.1 6973 UTR 3 XP_016869139.1

View Full Product Details