Product Details

SNP ID
rs117103320
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:42911728 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCATATTTTGATGGTTGGTATGT[A/G]GGGGAGTTGACTTTTAGGTTTTTAA
Phenotype
MIM: 607825
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HOOK3 PubMed Links

Gene Details

Gene
HOOK3
Gene Name
hook microtubule-tethering protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032410.3 Intron NP_115786.1

View Full Product Details