Product Details

SNP ID
rs117229732
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:122783444 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGAGGAGAGGATTCCTTCCCCCA[A/G]CACTTCTTGAATTGCCCAGACCTCC
Phenotype
MIM: 609185
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZHX2 PubMed Links

Gene Details

Gene
ZHX2
Gene Name
zinc fingers and homeoboxes 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014943.3 Intron NP_055758.1
XM_005250836.4 Intron XP_005250893.1
XM_005250837.4 Intron XP_005250894.1
XM_011516931.2 Intron XP_011515233.1
XM_011516932.2 Intron XP_011515234.1

View Full Product Details