Product Details

SNP ID
rs117531596
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:42175827 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGTCCTGAAAACTTCCAAAATGG[A/G]AAGTATCTGGGAAAATGCACTCTTC
Phenotype
MIM: 610469 MIM: 173370
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AP3M2 PubMed Links

Gene Details

Gene
AP3M2
Gene Name
adaptor related protein complex 3 mu 2 subunit
There are no transcripts associated with this gene.

Gene
PLAT
Gene Name
plasminogen activator, tissue type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000930.4 1974 UTR 3 NP_000921.1
NM_001319189.1 1974 UTR 3 NP_001306118.1
NM_033011.3 1974 UTR 3 NP_127509.1

View Full Product Details