Product Details

SNP ID
rs117665105
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:41931283 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGCTGGGTGGCGTGTGTGTGCGT[C/T]ATGGCTGATTCACCAGGTGGTAAAA
Phenotype
MIM: 601408
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KAT6A PubMed Links

Gene Details

Gene
KAT6A
Gene Name
lysine acetyltransferase 6A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305878.1 7593 Intron NP_001292807.1
NM_006766.4 7593 UTR 3 NP_006757.2
XM_011544656.2 7593 UTR 3 XP_011542958.1
XM_011544657.2 7593 UTR 3 XP_011542959.1
XM_011544658.2 7593 UTR 3 XP_011542960.1
XM_011544659.2 7593 UTR 3 XP_011542961.1
XM_017013863.1 7593 UTR 3 XP_016869352.1
XM_017013864.1 7593 UTR 3 XP_016869353.1

View Full Product Details