Product Details

SNP ID
rs121909356
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:96144951 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCCGCCGCCGGCGGCCGGGCGAG[T/G]GCAGCCAAGGCCTGGCATCCGGGGC
Phenotype
MIM: 601147
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
GDF6 PubMed Links

Gene Details

Gene
GDF6
Gene Name
growth differentiation factor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001557.2 1080 Missense Mutation CAC,CCC H327P NP_001001557.1

View Full Product Details