Product Details

SNP ID
rs138019004
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:67043260 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAGATGTTAATTTGATTAAACAG[C/T]TTATCCTTAATAACCTGAGACATCA
Phenotype
MIM: 604850
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COPS5 PubMed Links

Gene Details

Gene
COPS5
Gene Name
COP9 signalosome subunit 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006837.2 1309 Silent Mutation AAA,AAG K326K NP_006828.2

View Full Product Details