Product Details

SNP ID
rs138145251
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:124553152 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCCTGGGGAGACAGTGGCACTT[G/T]GGGGTTCCCGTTCCTGGTCTTCAGC
Phenotype
MIM: 608486 MIM: 601445
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MTSS1 PubMed Links

Gene Details

Gene
MTSS1
Gene Name
metastasis suppressor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282971.1 2210 Missense Mutation NP_001269900.1
NM_001282974.1 2210 Missense Mutation NP_001269903.1
NM_014751.5 2210 Missense Mutation NP_055566.3
XM_005251111.1 2210 Missense Mutation XP_005251168.1
XM_005251113.1 2210 Missense Mutation XP_005251170.1
XM_005251118.1 2210 Missense Mutation XP_005251175.1
XM_006716700.1 2210 Missense Mutation XP_006716763.1
XM_006716701.1 2210 Missense Mutation XP_006716764.1
XM_006716702.1 2210 Missense Mutation XP_006716765.1
XM_006716703.1 2210 Missense Mutation XP_006716766.1
XM_006716704.1 2210 Missense Mutation XP_006716767.1
XM_006716705.1 2210 Missense Mutation XP_006716768.1
XM_006716706.1 2210 Missense Mutation XP_006716769.1
XM_011517403.1 2210 Missense Mutation XP_011515705.1
XM_011517404.2 2210 Missense Mutation XP_011515706.1
XM_011517407.1 2210 Missense Mutation XP_011515709.1
XM_017014086.1 2210 Missense Mutation XP_016869575.1
XM_017014087.1 2210 Missense Mutation XP_016869576.1
XM_017014088.1 2210 Missense Mutation XP_016869577.1
XM_017014089.1 2210 Missense Mutation XP_016869578.1
XM_017014090.1 2210 Missense Mutation XP_016869579.1
XM_017014091.1 2210 Missense Mutation XP_016869580.1
XM_017014092.1 2210 Missense Mutation XP_016869581.1
XM_017014093.1 2210 Missense Mutation XP_016869582.1
XM_017014094.1 2210 Missense Mutation XP_016869583.1
XM_017014095.1 2210 Missense Mutation XP_016869584.1
XM_017014096.1 2210 Missense Mutation XP_016869585.1
XM_017014097.1 2210 Missense Mutation XP_016869586.1
XM_017014098.1 2210 Missense Mutation XP_016869587.1
XM_017014099.1 2210 Missense Mutation XP_016869588.1
XM_017014100.1 2210 Missense Mutation XP_016869589.1
XM_017014101.1 2210 Missense Mutation XP_016869590.1
Gene
NDUFB9
Gene Name
NADH:ubiquinone oxidoreductase subunit B9
There are no transcripts associated with this gene.

View Full Product Details