Product Details

SNP ID
rs138368338
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:135464801 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGAAAACCGAGGTCAGGAAGGTC[A/G]CATAATTTGCCTAAGAACACATAGC
Phenotype
MIM: 610421
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KHDRBS3 PubMed Links

Gene Details

Gene
KHDRBS3
Gene Name
KH RNA binding domain containing, signal transduction associated 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006558.2 Intron NP_006549.1
XM_005250757.2 Intron XP_005250814.1
XM_005250758.3 Intron XP_005250815.1
XM_011516796.2 Intron XP_011515098.1
XM_011516797.2 Intron XP_011515099.1
XM_011516798.1 Intron XP_011515100.1
XM_011516799.1 Intron XP_011515101.1

View Full Product Details