Product Details

SNP ID
rs138371466
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:22601006 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGAGGTGACAAGTTCAGAGGTGT[C/T]GGCAGGGAGGCACTCTTTCTCAAAC
Phenotype
MIM: 607359 MIM: 609722
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C8orf58 PubMed Links

Gene Details

Gene
C8orf58
Gene Name
chromosome 8 open reading frame 58
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013842.2 285 Silent Mutation GTC,GTT V55V NP_001013864.1
NM_001198827.1 285 Silent Mutation GTC,GTT V55V NP_001185756.1
NM_173686.2 285 Silent Mutation GTC,GTT V55V NP_775957.2
Gene
CCAR2
Gene Name
cell cycle and apoptosis regulator 2
There are no transcripts associated with this gene.

Gene
LOC107986876
Gene Name
proline-rich proteoglycan 2-like
There are no transcripts associated with this gene.

Gene
PDLIM2
Gene Name
PDZ and LIM domain 2
There are no transcripts associated with this gene.

View Full Product Details