Product Details

SNP ID
rs138669657
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:96262204 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTTCACCATCGTCAGCCTCATGT[A/T]CTTCGCCTTTACCAGGTGGGGCGGC
Phenotype
MIM: 616930 MIM: 612792
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MTERF3 PubMed Links

Gene Details

Gene
MTERF3
Gene Name
mitochondrial transcription termination factor 3
There are no transcripts associated with this gene.

Gene
PTDSS1
Gene Name
phosphatidylserine synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290225.1 319 UTR 5 NP_001277154.1
NM_014754.2 319 Missense Mutation TAC,TTC Y55F NP_055569.1

View Full Product Details