Product Details
- SNP ID
-
rs139278478
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:116771616 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCTCAGTGCATGAGAAAGAAAGTA[C/T]CAAACATCTCAAAGAGGAACAGGGT
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
UTP23
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1133950] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- UTP23
- Gene Name
- UTP23, small subunit processome component
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032334.2 |
625 |
Missense Mutation |
ACC,ATC |
T175I |
NP_115710.2 |
View Full Product Details