Product Details

SNP ID
rs139316534
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143366780 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAAGAAGCCTGCAAAACAAATAC[A/G]TAAGAACTGGATACCATTTCAGTCA
Phenotype
MIM: 601031 MIM: 606387
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RHPN1 PubMed Links

Gene Details

Gene
RHPN1
Gene Name
rhophilin Rho GTPase binding protein 1
There are no transcripts associated with this gene.

Gene
RHPN1-AS1
Gene Name
RHPN1 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
TOP1MT
Gene Name
topoisomerase (DNA) I, mitochondrial
There are no transcripts associated with this gene.

View Full Product Details