Product Details
- SNP ID
-
rs139553486
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:119557530 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAGATGCTCAGAAAGTTAAATCTC[A/G]CTCTCATATGTATGCAGGTATGTCT
- Phenotype
-
MIM: 601060
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ENPP2
PubMed Links
Gene Details
- Gene
- ENPP2
- Gene Name
- ectonucleotide pyrophosphatase/phosphodiesterase 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001040092.2 |
2669 |
Silent Mutation |
AGC,AGT |
S861S |
NP_001035181.1 |
NM_001130863.2 |
2669 |
Silent Mutation |
AGC,AGT |
S886S |
NP_001124335.1 |
NM_006209.4 |
2669 |
Silent Mutation |
AGC,AGT |
S913S |
NP_006200.3 |
XM_006716584.1 |
2669 |
Silent Mutation |
AGC,AGT |
S938S |
XP_006716647.1 |
XM_006716585.1 |
2669 |
Silent Mutation |
AGC,AGT |
S934S |
XP_006716648.1 |
XM_006716587.1 |
2669 |
Silent Mutation |
AGC,AGT |
S909S |
XP_006716650.1 |
XM_017013571.1 |
2669 |
Silent Mutation |
AGC,AGT |
S882S |
XP_016869060.1 |
XM_017013572.1 |
2669 |
Silent Mutation |
AGC,AGT |
S882S |
XP_016869061.1 |
XM_017013573.1 |
2669 |
Silent Mutation |
AGC,AGT |
S878S |
XP_016869062.1 |
XM_017013574.1 |
2669 |
Silent Mutation |
AGC,AGT |
S857S |
XP_016869063.1 |
XM_017013575.1 |
2669 |
Silent Mutation |
AGC,AGT |
S857S |
XP_016869064.1 |
View Full Product Details