Product Details

SNP ID
rs139651821
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:66429978 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGAACAGCAAGAAGCCTCAGCTG[C/G]ATGTGACTAGGGCCACCAATAAGCA
Phenotype
MIM: 611083
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ADHFE1 PubMed Links

Gene Details

Gene
ADHFE1
Gene Name
alcohol dehydrogenase, iron containing 1
There are no transcripts associated with this gene.

Gene
RRS1
Gene Name
ribosome biogenesis regulator homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015169.3 951 Missense Mutation CAT,GAT H283D NP_055984.1
Gene
RRS1-AS1
Gene Name
RRS1 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details