Product Details

SNP ID
rs139681634
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143158313 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGAGCCCCCCGGCCAGGGCCCA[C/G]GGGCTGTGCCCTGCCCCTGCCGCCC
Phenotype
MIM: 603625
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LY6H PubMed Links

Gene Details

Gene
LY6H
Gene Name
lymphocyte antigen 6 complex, locus H
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130478.1 526 Silent Mutation CCC,CCG P141P NP_001123950.1
NM_001135655.1 526 Silent Mutation CCC,CCG P141P NP_001129127.1
NM_002347.4 526 Silent Mutation CCC,CCG P120P NP_002338.3
XM_017013413.1 526 Silent Mutation CCC,CCG P146P XP_016868902.1
XM_017013414.1 526 Silent Mutation CCC,CCG P120P XP_016868903.1

View Full Product Details