Product Details

SNP ID
rs139946542
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:123684670 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCATCGGTCCCCGCACCCTTCAT[C/T]GACTTGTACAGACGTTCAGCAAAAT
Phenotype
MIM: 602573
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANXA13 PubMed Links

Gene Details

Gene
ANXA13
Gene Name
annexin A13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003954.2 863 Silent Mutation TCA,TCG S298S NP_001003954.1
NM_004306.3 863 Silent Mutation TCA,TCG S257S NP_004297.2

View Full Product Details