Product Details

SNP ID
rs140003920
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144359876 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGGCACTGGCATGCTGTGCCTC[A/G]AATGTCACTTTCCTGCCCTTCTTGA
Phenotype
MIM: 609076 MIM: 607882
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXL6 PubMed Links

Gene Details

Gene
FBXL6
Gene Name
F-box and leucine rich repeat protein 6
There are no transcripts associated with this gene.

Gene
LOC101928902
Gene Name
uncharacterized LOC101928902
There are no transcripts associated with this gene.

Gene
SLC52A2
Gene Name
solute carrier family 52 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253815.1 945 Silent Mutation TCA,TCG S128S NP_001240744.1
NM_001253816.1 945 Silent Mutation TCA,TCG S128S NP_001240745.1
NM_024531.4 945 Silent Mutation TCA,TCG S128S NP_078807.1
XM_006716658.2 945 Silent Mutation TCA,TCG S128S XP_006716721.1
XM_006716659.2 945 Silent Mutation TCA,TCG S128S XP_006716722.1
XM_006716660.2 945 Silent Mutation TCA,TCG S128S XP_006716723.1
XM_011517300.2 945 Silent Mutation TCA,TCG S40S XP_011515602.1
XM_017013819.1 945 Silent Mutation TCA,TCG S128S XP_016869308.1
XM_017013820.1 945 Silent Mutation TCA,TCG S128S XP_016869309.1
XM_017013821.1 945 Intron XP_016869310.1
XM_017013822.1 945 Intron XP_016869311.1
Gene
TMEM249
Gene Name
transmembrane protein 249
There are no transcripts associated with this gene.

View Full Product Details