Product Details

SNP ID
rs140042949
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144105966 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCACGTGCTGGAGGCACTTTCT[C/T]CACCCCAGACTTCAGGACTGAGCCC
Phenotype
MIM: 123980 MIM: 610210 MIM: 611885
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYC1 PubMed Links

Gene Details

Gene
CYC1
Gene Name
cytochrome c1
There are no transcripts associated with this gene.

Gene
MAF1
Gene Name
MAF1 homolog, negative regulator of RNA polymerase III
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032272.4 805 Missense Mutation CCA,TCA P61S NP_115648.2
XM_017013903.1 805 Missense Mutation CCA,TCA P61S XP_016869392.1
XM_017013904.1 805 Intron XP_016869393.1
Gene
SHARPIN
Gene Name
SHANK associated RH domain interactor
There are no transcripts associated with this gene.

Gene
WDR97
Gene Name
WD repeat domain 97
There are no transcripts associated with this gene.

View Full Product Details