Product Details

SNP ID
rs140164489
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:43103599 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGAGGCCTCGCCCCCCGAGAGGT[G/T]CCGCCAGCTGTTGGGCTGCTGCTGA
Phenotype
MIM: 615247
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
POMK PubMed Links

Gene Details

Gene
POMK
Gene Name
protein-O-mannose kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277971.1 222 Silent Mutation GTG,GTT V17V NP_001264900.1
NM_032237.4 222 Silent Mutation GTG,GTT V17V NP_115613.1

View Full Product Details