Product Details

SNP ID
rs140227767
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:6808641 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTGAGGAAGCCATCTGAGAAGCC[A/G]ATGTTGGTCCACAGGGCCTCGAACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
XKR5 PubMed Links

Gene Details

Gene
XKR5
Gene Name
XK related 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289973.1 4907 UTR 3 NP_001276902.1
NM_207411.4 4907 UTR 3 NP_997294.3
XM_011534750.2 4907 Intron XP_011533052.1
XM_017013401.1 4907 Intron XP_016868890.1
XM_017013402.1 4907 Intron XP_016868891.1

View Full Product Details