Product Details

SNP ID
rs140910313
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:30132001 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAAGCAGAATACAGTACACCATG[G/A]GAAAGACACTGTTGTACGAATTACA
Phenotype
MIM: 607338 MIM: 611940
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
LEPROTL1 PubMed Links

Gene Details

Gene
LEPROTL1
Gene Name
leptin receptor overlapping transcript-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128208.1 1333 Intron NP_001121680.1
NM_015344.2 1333 Intron NP_056159.2
Gene
MBOAT4
Gene Name
membrane bound O-acyltransferase domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100916.1 1333 Missense Mutation CCC,CTC P417L NP_001094386.1

View Full Product Details