Product Details

SNP ID
rs140936906
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143837696 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCGAAGAGCACGCGGTGGAAGAG[A/G]AGGCTGTGGGCAGAGGGCCGGCGGG
Phenotype
MIM: 615563 MIM: 604819
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR6845 PubMed Links

Gene Details

Gene
MIR6845
Gene Name
microRNA 6845
There are no transcripts associated with this gene.

Gene
NRBP2
Gene Name
nuclear receptor binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178564.3 1040 Silent Mutation CTC,CTT L300L NP_848659.2
XM_017013378.1 1040 Silent Mutation CTC,CTT L300L XP_016868867.1
XM_017013379.1 1040 Silent Mutation CTC,CTT L300L XP_016868868.1
XM_017013380.1 1040 Silent Mutation CTC,CTT L300L XP_016868869.1
XM_017013381.1 1040 Silent Mutation CTC,CTT L300L XP_016868870.1
XM_017013382.1 1040 Silent Mutation CTC,CTT L300L XP_016868871.1
XM_017013383.1 1040 Silent Mutation CTC,CTT L300L XP_016868872.1
Gene
PUF60
Gene Name
poly(U) binding splicing factor 60
There are no transcripts associated with this gene.

View Full Product Details