Product Details

SNP ID
rs141302014
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143559365 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCGGCCTTTGAGCGGGTAGTCCG[A/G]AGAGTGGTCCAGGAGCTGGACCATG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GSDMD PubMed Links

Gene Details

Gene
GSDMD
Gene Name
gasdermin D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166237.1 733 Silent Mutation CGA,CGG R10R NP_001159709.1
NM_024736.6 733 Silent Mutation CGA,CGG R10R NP_079012.3
XM_011517301.1 733 Silent Mutation CGA,CGG R10R XP_011515603.1
Gene
MROH6
Gene Name
maestro heat like repeat family member 6
There are no transcripts associated with this gene.

Gene
ZC3H3
Gene Name
zinc finger CCCH-type containing 3
There are no transcripts associated with this gene.

View Full Product Details