Product Details

SNP ID
rs141530073
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:81442119 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAATACATATAAACGTACATATAC[A/G]TGGTTTAGAAATTATGAATTTATAC
Phenotype
MIM: 170715
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PMP2 PubMed Links

Gene Details

Gene
PMP2
Gene Name
peripheral myelin protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002677.3 1776 UTR 3 NP_002668.1

View Full Product Details