Product Details

SNP ID
rs141540799
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:142701602 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTTTTTATTCCTCCTTTCAGAC[A/G]AGGGTGACAATAGAGTGTGGTGTCA
Phenotype
MIM: 615093
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100288181 PubMed Links

Gene Details

Gene
LOC100288181
Gene Name
uncharacterized LOC100288181
There are no transcripts associated with this gene.

Gene
LY6K
Gene Name
lymphocyte antigen 6 complex, locus K
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160354.1 711 Missense Mutation AAG,GAG K36E NP_001153826.1
NM_001160355.1 711 Missense Mutation AAG,GAG K36E NP_001153827.1
NM_017527.3 711 Missense Mutation AAG,GAG K36E NP_059997.3
XM_005250989.4 711 Missense Mutation AAG,GAG K94E XP_005251046.1
XM_005250990.4 711 Missense Mutation AAG,GAG K94E XP_005251047.1

View Full Product Details