Product Details

SNP ID
rs141703643
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143440304 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGGGAGGTGCAGCCACGGCAGCC[A/G]CAGTGAGGGCAGCCGAGCTGGGCGT
Phenotype
MIM: 610303
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAFA PubMed Links

Gene Details

Gene
MAFA
Gene Name
MAF bZIP transcription factor A
There are no transcripts associated with this gene.

Gene
ZC3H3
Gene Name
zinc finger CCCH-type containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015117.2 2581 Missense Mutation GCG,GTG A851V NP_055932.2
XM_006716536.3 2581 Missense Mutation GCG,GTG A913V XP_006716599.2
XM_011516943.2 2581 Intron XP_011515245.1
XM_011516944.2 2581 Intron XP_011515246.1
XM_017013248.1 2581 Missense Mutation GCG,GTG A979V XP_016868737.1
XM_017013249.1 2581 Missense Mutation GCG,GTG A979V XP_016868738.1

View Full Product Details