Product Details

SNP ID
rs141757417
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:133456733 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAGAAATGCAGTCTCCTGCACGG[C/G]AGGGTGTGAGAAGGCCACCCAGGCA
Phenotype
MIM: 607187
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ST3GAL1 PubMed Links

Gene Details

Gene
ST3GAL1
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003033.3 5342 Intron NP_003024.1
NM_173344.2 5342 Intron NP_775479.1
XM_005251025.4 5342 UTR 3 XP_005251082.1
XM_006716617.1 5342 UTR 3 XP_006716680.1
XM_011517225.1 5342 UTR 3 XP_011515527.1
XM_017013736.1 5342 UTR 3 XP_016869225.1
XM_017013737.1 5342 UTR 3 XP_016869226.1

View Full Product Details