Product Details

SNP ID
rs141879192
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:120396121 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATTCTCTTATAGCCGATAATCTT[C/T]AGGTCTGAAAGAAAAAATCAACATA
Phenotype
MIM: 610200
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRPL13 PubMed Links

Gene Details

Gene
MRPL13
Gene Name
mitochondrial ribosomal protein L13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014078.5 833 Missense Mutation AAA,GAA K174E NP_054797.2

View Full Product Details