Product Details

SNP ID
rs142005076
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:126556841 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCACCCACGTGCGCCAGCGCGTT[A/G]CGCACCACGGCGCTGGAGCTTAGCG
Phenotype
MIM: 609483
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM84B PubMed Links

Gene Details

Gene
FAM84B
Gene Name
family with sequence similarity 84 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174911.4 1078 Silent Mutation CGC,CGT R183R NP_777571.1
XM_017013107.1 1078 Silent Mutation CGC,CGT R183R XP_016868596.1
XM_017013108.1 1078 Silent Mutation CGC,CGT R165R XP_016868597.1

View Full Product Details