Product Details
- SNP ID
-
rs142101522
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:132710899 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAATAAAACACTTGATTCCAAGTAG[A/T]CTGCAAGTTGGACAATTTCCAGATA
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM71
PubMed Links
Gene Details
- Gene
- TMEM71
- Gene Name
- transmembrane protein 71
View Full Product Details