Product Details

SNP ID
rs142256352
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:11339645 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTGTTCCCTTACAGAAAATTTGC[C/T]GACCTCTGCTCTGGAACGTCAGGCT
Phenotype
MIM: 615199 MIM: 615174
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC35G5 PubMed Links

Gene Details

Gene
SLC35G5
Gene Name
solute carrier family 35 member G5
There are no transcripts associated with this gene.

Gene
TDH
Gene Name
L-threonine dehydrogenase (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details