Product Details

SNP ID
rs142387618
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144791299 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCAACCACAGCTCTGTTGGCTGA[A/G]GAGATAACCTTCTTGGAGCCGGAGG
Phenotype
MIM: 604177 MIM: 194526
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR6850 PubMed Links

Gene Details

Gene
MIR6850
Gene Name
microRNA 6850
There are no transcripts associated with this gene.

Gene
RPL8
Gene Name
ribosomal protein L8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000973.4 612 Silent Mutation NP_000964.1
NM_001317771.1 612 Silent Mutation NP_001304700.1
NM_001317782.1 612 Silent Mutation NP_001304711.1
NM_033301.2 612 Silent Mutation NP_150644.1
XM_017013723.1 612 Silent Mutation XP_016869212.1
Gene
ZNF34
Gene Name
zinc finger protein 34
There are no transcripts associated with this gene.

Gene
ZNF517
Gene Name
zinc finger protein 517
There are no transcripts associated with this gene.

View Full Product Details