Product Details

SNP ID
rs142606096
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:94250346 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGTCATGGCAGGATTTGGACTT[C/G]AGCTTGAAGGCCATATTCTTGTTGT
Phenotype
MIM: 600164
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GEM PubMed Links

Gene Details

Gene
GEM
Gene Name
GTP binding protein overexpressed in skeletal muscle
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005261.3 1298 Silent Mutation CTC,CTG L285L NP_005252.1
NM_181702.2 1298 Silent Mutation CTC,CTG L285L NP_859053.1
XM_017013315.1 1298 UTR 3 XP_016868804.1

View Full Product Details