Product Details

SNP ID
rs142687555
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:79764337 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGACCACTCGCACACCATGATCA[C/G]TTATCCATGTTAACATTTGTGAATT
Phenotype
MIM: 602953
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HEY1 PubMed Links

Gene Details

Gene
HEY1
Gene Name
hes related family bHLH transcription factor with YRPW motif 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040708.1 1708 UTR 3 NP_001035798.1
NM_001282851.1 1708 UTR 3 NP_001269780.1
NM_012258.3 1708 UTR 3 NP_036390.3
Gene
LINC01607
Gene Name
long intergenic non-protein coding RNA 1607
There are no transcripts associated with this gene.

Gene
LOC101927040
Gene Name
uncharacterized LOC101927040
There are no transcripts associated with this gene.

View Full Product Details