Product Details

SNP ID
rs142805240
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143568162 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGACCAGGCGGCAGCAGAGGTGGC[C/T]CAGGGCCTCGGGGCTGTCATAGTGG
Phenotype
MIM: 611552
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GSDMD PubMed Links

Gene Details

Gene
GSDMD
Gene Name
gasdermin D
There are no transcripts associated with this gene.

Gene
MROH6
Gene Name
maestro heat like repeat family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100878.1 1785 Missense Mutation AGC,GGC S582G NP_001094348.1
XM_006716615.2 1785 UTR 3 XP_006716678.1
XM_011517214.1 1785 Missense Mutation AGC,GGC S583G XP_011515516.1
XM_011517215.1 1785 Missense Mutation AGC,GGC S582G XP_011515517.1
XM_011517216.2 1785 Missense Mutation AGC,GGC S583G XP_011515518.1
XM_011517217.1 1785 Missense Mutation AGC,GGC S583G XP_011515519.1
XM_011517218.1 1785 Intron XP_011515520.1
XM_011517221.2 1785 Missense Mutation AGC,GGC S583G XP_011515523.1
XM_011517222.1 1785 Missense Mutation AGC,GGC S583G XP_011515524.1
XM_011517223.1 1785 Missense Mutation AGC,GGC S583G XP_011515525.1
Gene
NAPRT
Gene Name
nicotinate phosphoribosyltransferase
There are no transcripts associated with this gene.

View Full Product Details