Product Details
- SNP ID
-
rs142854962
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:11995233 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAATATTGTACACTATATGGGGTT[A/C]TTTGTCTGGAGGCCACAAATCCATT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DEFB134
PubMed Links
Gene Details
- Gene
- DEFB134
- Gene Name
- defensin beta 134
View Full Product Details