Product Details

SNP ID
rs143309476
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:116766632 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGATCACAAGGCAGAAACATGCCA[A/G]GAAGCATCTTGGCTTCTTCCGCAAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UTP23 PubMed Links

Gene Details

Gene
UTP23
Gene Name
UTP23, small subunit processome component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032334.2 130 Missense Mutation AAG,AGG K10R NP_115710.2

View Full Product Details