Product Details

SNP ID
rs143401399
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:10525595 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCACAGCCCAGGGCCATGCTCCTG[G/T]TCTCAGTGTTGCTGCTCCTGTCCCT
Phenotype
MIM: 615144
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PRSS55 PubMed Links

Gene Details

Gene
PRSS55
Gene Name
protease, serine 55
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197020.1 265 Missense Mutation GTC,TTC V4F NP_001183949.1
NM_198464.3 265 Missense Mutation GTC,TTC V4F NP_940866.2
XM_005272372.3 265 Missense Mutation GTC,TTC V69F XP_005272429.2
XM_011543814.2 265 Missense Mutation GTC,TTC V69F XP_011542116.2
XM_017013182.1 265 Missense Mutation GTC,TTC V69F XP_016868671.1

View Full Product Details