Product Details

SNP ID
rs143532856
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:22138644 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCGTCCTCCCACACTGACCTCCC[A/G]CACCGGTGGCTTCCTCTTGACCACA
Phenotype
MIM: 602302 MIM: 608302 MIM: 609349
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HR PubMed Links

Gene Details

Gene
HR
Gene Name
hair growth associated
There are no transcripts associated with this gene.

Gene
LGI3
Gene Name
leucine rich repeat LGI family member 3
There are no transcripts associated with this gene.

Gene
REEP4
Gene Name
receptor accessory protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316964.1 1172 Intron NP_001303893.1
NM_001316965.1 1172 UTR 3 NP_001303894.1
NM_025232.3 1172 Missense Mutation CGG,TGG R235W NP_079508.2

View Full Product Details