Product Details

SNP ID
rs143559181
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:72023115 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGGAGAAAAGTAAGATCCTTCA[G/T]CCTAAAAGGACATACACATTTCATG
Phenotype
MIM: 604775
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MSC-AS1 PubMed Links

Gene Details

Gene
MSC-AS1
Gene Name
MSC antisense RNA 1
There are no transcripts associated with this gene.

Gene
TRPA1
Gene Name
transient receptor potential cation channel subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007332.2 3326 Missense Mutation ATG,CTG M1051L NP_015628.2
XM_011517624.2 3326 Missense Mutation ATG,CTG M1076L XP_011515926.1
XM_011517625.2 3326 Missense Mutation ATG,CTG M1051L XP_011515927.1
XM_017013946.1 3326 Missense Mutation ATG,CTG M1051L XP_016869435.1

View Full Product Details